Lutetium 1000 mg/L Lu in 5% salpetersyre (fra Lu₂O₃) Specpure® standard til AAS

Leverandør: Thermo Fisher Scientific

89886.AE 89886.AP
89886.AEEA 1025 NOK
89886.AE 89886.AP
Lutetium 1000 mg/L Lu in 5% salpetersyre (fra Lu₂O₃) Specpure® standard til AAS
Standarder Kationstandarder Lutetium Standards

Molecular Formula: Lu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR