Tantalum 1000 mg/L Ta in 2% hydrofluoric acid (fra TaCl₅) Specpure® standard til AAS

Leverandør: Thermo Fisher Scientific

89888.AE 89888.AP
89888.AEEA 555 NOK
89888.AE 89888.AP
Tantalum 1000 mg/L Ta in 2% hydrofluoric acid (fra TaCl₅) Specpure® standard til AAS
Standarder Kationstandarder Tantalum Standards

MDL No.: MFCD00011252
Molecular Formula: TaCl₅ in 2% HF
UN No.: 2922
ADR: 8,II
GHS Symbol(s): GHS06-GHS05
GHS Signal Word: Danger


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR