EDTA dinatrium salt dihydrat 99.0-101.0%, krystaller, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor

8993-01EA 4050 NOK
8993-01 8993-05 8993-R 8993-07
EDTA dinatrium salt dihydrat 99.0-101.0%, krystaller, BAKER ANALYZED® ACS, J.T.Baker®
EDTA dinatrium salt dihydrat

The USP/NF Offical Compendia of Standards contains a section titled, "Reagents, Indicators, and Solutions" that lists the quality requirements of chemicals to be used in compendia testing. PharmaTest reagents are tested according to these requirements, relieving you of the need to do additional testing on your reagents. Products may be reagent or other grade, but are labeled as "Meets reagent specifications for testing USP/NF monographs." Test solutions are labeled as PharmaTest.

Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 255 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 101.0 %
Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O) 99.0 - 101.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 4.0 - 6.0
Nitrilotriacetic Acid (by polarography) ≤0.1 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.01 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR