EDTA dinatrium salt dihydrat 99.0-101.0% (dried basis), krystaller USP, Multi-Compendial, J.T.Baker®

Leverandør: Avantor

8995-01EA 3375 NOK
8995-01 8995-07 8995-20
EDTA dinatrium salt dihydrat 99.0-101.0% (dried basis), krystaller USP, Multi-Compendial, J.T.Baker®
EDTA dinatrium salt dihydrat

GMP Manufactured Product Meets B.P. Chemical Specifications Meets E.P. Chemical Specifications Meets J.P. Chemical Specifications Meets U.S.P Requirements

Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 255 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - pH (1 in 20) 4.0 - 6.0
USP - Loss on Drying 8.7 - 11.4 %
USP - Calcium (Ca) Passes Test
USP - Nitrilotriacetic Acid ≤0.1 %
USP - Assay (C₁₀H₁₄N₂Na₂O₈) (dried basis) 99.0 - 101.0 %
Endotoxin Concentration (EU/g) ≤5
EP/BP - Assay 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification D Passes Test
EP/BP - Impurity A ≤0.1 %
EP/BP - Appearance of Solution Passes Test
EP/BP - pH (1 in 20) 4.0 - 5.5
EP/BP - Iron (Fe) ≤80 ppm
JP - Assay 99.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Identification C Passes Test
JP - pH of 1% Solution at 25°C 4.3 - 4.7
JP - Clarity and Color of Solution Passes Test
JP - Cyanide (CN) Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Residue after Ignition 37.0 - 39.0 %
No Class 1,2,3 or other solvents are used or produced in the 37.0 - 39.0 %
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR