FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥98% (by TLC) amine-reactive fluorescent dye

Leverandør: Biotium

Synonymer: 5-isothiocyanatofluorescein, Fluorescein 5(6)-isothiocyanate, Fluorescein Isothiocyanate, Isomer I

90020
90020.EA 2425 NOK
90020.
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥98% (by TLC) amine-reactive fluorescent dye
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)

FITC (fluorescein-5-isothiocyanate) is an amine-reactive fluorescein derivative for labeling proteins or other molecules containing primary amines.


  • λEx/λEm= 492/520 nm
  • ε = 75,000
  • Orange solid soluble in DMF or DMSO
  • Store at -20°C and protect from light, especially in solution

Formel: C₂₁H₁₁NO₅S
MW: 389.381 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00005063
CAS-nummer: 3326-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5c22245563f0151df608ac5268e64ceb733e596&vid=a5c22245563f0151df608ac5268e64ceb733e596&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR