5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥98% (by TLC) amine-reactive fluorescent dye

Leverandør: Biotium

Synonymer: (5(6)-TAMRA SE

90022
90022.EA 3500 NOK
90022.
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] ≥98% (by TLC) amine-reactive fluorescent dye
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

5(6)-TAMRA SE (full name: 5-(and-6)-Carboxytetramethylrhodamine succinimidyl ester (mixed isomers)) is the amine reactive form of TAMRA, one of the most commonly used red fluorescent dyes. The succinimidyl ester reacts readily with primary or secondary amines under mild conditions.


  • λEx/λEm (MeOH) = 540/565 nm
  • ε = 95,000
  • Red solid soluble in DMF or DMSO
  • Store at -20°C and protect from light

Formel: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00077324
CAS-nummer: 150408-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d953e7201678452a2b20e33e8865d0160fc6f83e&vid=d953e7201678452a2b20e33e8865d0160fc6f83e&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR