Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Leverandør: Merck
90568-10MG
90568-10MGEA 995 NOK
90568-10MG
Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Standarder Organiske standarder
Used as a reference standard in determination of fenofibric acid in samples of human plasma using high performance liquid chromatography (HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce64e9c732507d863c7f425cdbc3aa408ad045b7&vid=ce64e9c732507d863c7f425cdbc3aa408ad045b7&prid=web&clid=SAPDAM