Tetrabutylammoniumjodid ≥99%

Leverandør: VWR Chemicals
Warning

Synonymer: Tetra-n-butylammonium iodide, TBAI

91244.180EA 620 NOK
91244.180
Tetrabutylammoniumjodid ≥99%
Tetrabutylammoniumjodid
Formel: C₁₆H₃₆IN
Smeltepunkt: 143…146 °C
Tetthet: 1,0027 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011636
CAS-nummer: 311-28-4
EINECS: 206-220-5

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR