Ammoniumdihydrogenortofosfat 100 g/L in nitric acid 1%, ARISTAR® matrix modifier solution, for graphite furnace AAS

Leverandør: VWR Chemicals
Warning

91275.154EA 1500 NOK
91275.154
Ammoniumdihydrogenortofosfat 100 g/L in nitric acid 1%, ARISTAR® matrix modifier solution, for graphite furnace AAS
Ammoniumdihydrogenortofosfat

The VWR® Matrix modifier solutions have especially been developed for use in GF-AAS to eliminate matrix effects. They can be used either diluted or as mixtures.


  • Guaranteed accuracy: ±1%
  • COA incl. up to 70 measured trace impurities in ppm range


VWR® Matrix modifier solutions are produced with high purity metals or salts (≥99,999%) in sub-boiling distilled acids and are supplied with a detailed batch specific certificate of analysis.

Formel: NH₄H₂PO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003396
CAS-nummer: 7722-76-1
FN: 1760
ADR: 8,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR