VWR®, Mixed Anions Standard, 7 components in H₂O
Leverandør: VWR Chemicals
91295.180EA 2350 NOK
91295.180
VWR®, Mixed Anions Standard, 7 components in H₂O
Standarder Anionstandarder Multi-Element Anioinic Standards
7 components: 100 mg/l each of Chlorides (Cl-), Fluorides (F-), Sulphates (SO42-), Nitrates (NO3-), Nitrites (NO2-), Bromides (Br-), Phosphates (PO43-) in H₂O
Sertifiseringer: ISO 17025/17034
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp Ambient