VWR®, Mixed Anions Standard, 7 components in H₂O
Leverandør: VWR Chemicals
91296.180EA 2200 NOK
91296.180
VWR®, Mixed Anions Standard, 7 components in H₂O
Standarder Anionstandarder Multi-Element Anioinic Standards
7 components: Fluorides (F-) 2ppm, Phosphates (PO43-) 10ppm, Nitrites (NO2-) 5ppm, Nitrates (NO3-) 10ppm, Bromides (Br-) 10ppm, Chlorides (Cl-) 5ppm, Sulphates (SO42-) 10ppm in H₂O
Sertifiseringer: ISO 17025/17034
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp Ambient