VWR®, FAME test method for diesel and petro/biodiesel (6 comp, in n-heptane), CRM
Leverandør: VWR Chemicals
91681.050EA 3075 NOK
91681.050
VWR®, FAME test method for diesel and petro/biodiesel (6 comp, in n-heptane), CRM
Standarder PetroleumIndustristandarder
At stated conc. (µg/ml): Palmitic acid methyl ester (800); Stearic acid methyl ester (800); Methyl cis-9-octadecenoate (800); Linoleic acid methyl ester (800); Chrysene (400); Linolenic acid methyl ester (800).
Sertifiseringer: Certified reference material.
NB: Highly Flammable.