Supelco® TraceCERT® Organic Reference Standard, Biotin
Leverandør: Merck
91827-50MG
91827-50MGEA 870 NOK
91827-50MG
Supelco® TraceCERT® Organic Reference Standard, Biotin
Standarder Organiske standarder
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2020734fee2c4689f6d35c590644063a7d29b70f&vid=2020734fee2c4689f6d35c590644063a7d29b70f&prid=web&clid=SAPDAM