Supelco® TraceCERT® Organic Reference Standard, Dibenz[a,h]anthracene
Leverandør: Merck
91861-10MG
91861-10MGEA 885 NOK
91861-10MG
Supelco® TraceCERT® Organic Reference Standard, Dibenz[a,h]anthracene
Standarder Organiske standarder
  • GHS signal: GHS08, GHS09
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
NB: P201 - P273 - P308 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29932b8e7b22c704495f0b9c8b53ce8b1669069a&vid=29932b8e7b22c704495f0b9c8b53ce8b1669069a&prid=web&clid=SAPDAM