Organic reference standard, Phloretin, analytical standard, Supelco®
Leverandør: Merck
91897-10MG
91897-10MGEA 775 NOK
91897-10MG
Organic reference standard, Phloretin, analytical standard, Supelco®
Standarder Organiske standarder
Major polyphenol found in apple and aglycone of phloridzin.
Reacts with vic-dicarbonyl compounds such as glyoxal and methylglyoxal, preventing cytotoxic conjugation with biological macromolecules.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a96aa544de24fd1692ef06c737cfa95d856290&vid=86a96aa544de24fd1692ef06c737cfa95d856290&prid=web&clid=SAPDAM