Supelco® TraceCERT® Organic Reference Standard, Potassium phosphate monobasic

Leverandør: Merck

92214-1G
92214-1GEA 1400 NOK
92214-1G
Supelco® TraceCERT® Organic Reference Standard, Potassium phosphate monobasic
Standarder Organiske standarder

Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.

Bestill nå

SPESIFIKASJONER

Synonym(s) Monopotassium phosphate; Potassium dihydrogen phosphate; prim.-Potassium phosphate
Environmentally Preferable
M-Clarity quality level MQ300
Tetthet 2,338 g/ml at 25 °C (lit.)
CAS-nr 7778-77-0
MDL No. MFCD00011401
Formel KH₂PO₄
Molekylvekt 136,09
Smeltepunkt 252.6 °C (lit.)
Fraktforhold STD
Lagringsforhold 15...25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR