VWR®, Bisphenol A bis(diphenyl phosphate) (BADP, BAPP, BPADP, BDP), 100 µg/ml in toluene, CRM

Leverandør: VWR Chemicals

92340.010EA 530 NOK
92340.010
VWR®, Bisphenol A bis(diphenyl phosphate) (BADP, BAPP, BPADP, BDP), 100 µg/ml in toluene, CRM
Standarder Organiske standarder

Sertifiseringer: Certified reference material.


NB: Highly Flammable.

Bestill nå

SPESIFIKASJONER

Beskrivelse Bisphenol A bis(diphenyl phosphate) (BADP,BAPP,BPADP,BDP)
Environmentally Preferable
CAS-nr 5945-33-5
UN No. 1294
Lagringsforhold Ambient (>5 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR