Kloroform ≥99.8% (by GC, corrected for water content) stabilised, ULTRA RESI-ANALYZED® for analyse av organiskrester, J.T.Baker®

Leverandør: Avantor
Danger


Stabilisering: stabilisert med etanol ~0,75%
Packaged under inert gas.

9257-03EA 4425 NOK
9257-03 9257-02
Kloroform ≥99.8% (by GC, corrected for water content) stabilised, ULTRA RESI-ANALYZED® for analyse av organiskrester, J.T.Baker®
Kloroform
Formel: CHCl₃
Kokepunkt: 61,7 °C (1013 hPa)
Smeltepunkt: –63 °C
Tetthet: 1,4832 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000826
CAS-nummer: 67-66-3
EINECS: 200-663-8
FN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Indeks: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 10
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Chloride (Cl) ≤10 ppm
Color (APHA) ≤10
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Titrable Acid (µeq/g) ≤0.5
Water (by KF, coulometric) ≤0.05 %

SPESIFIKASJONER

Environmentally Preferable
Miljøsertifikat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR