Sykloheksan ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: Hexahydrobenzene, Hexamethylene

Packaged under inert gas.

9258-03EA 0 NOK
9258-03
Sykloheksan ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Sykloheksan
Formel: C₆H₁₂
MW: 84.16 g/mol
Kokepunkt: 81
Smeltepunkt: 7
Tetthet: 0.7785 g/cm³ (20 °C)
Flash punkt: –18 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003814
CAS-nummer: 110-82-7
EINECS: 203-806-2
FN: 1145
ADR: 3 II
Merck Indeks: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 10
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (C₆H₁₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤10
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR