Sykloheksan, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Leverandør: Avantor

9258.1000EA 1350 NOK
9258.1000 9258.2500
Sykloheksan, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Sykloheksan
Formel: C₆H₁₂
MDL nummer: MFCD00003814
CAS-nummer: 110-82-7
FN: 1145
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (by GC) (corrected for water) min. 99.0 %
Color (APHA) max. 10
Residue after Evaporation max. 2 ppm
Substances Darkened by H2SO4 passes test
Water (by KF, coulometric) max. 100 ppm
ECD Sensitive Impurities (as Heptachlor Epoxide)
Single Impurity Peak (pg/ml) max. 10
FID-Sensitive Impurities (as 2-Octanol)
Single Impurity Peak (ng/ml) max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR