Etylacetat ≥99.6% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Leverandør: Avantor
Danger


Packaged under inert gas.

9260.1000EA 1675 NOK
9260.1000 9260.2500 9260-03
Etylacetat ≥99.6% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Etylacetat
Formel: CH₃COOC₂H₅
Kokepunkt: 77,1 °C (1013 hPa)
Smeltepunkt: –83 °C
Tetthet: 0,902 g/cm³ (20 °C)
Flash punkt: –4 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009171
CAS-nummer: 141-78-6
EINECS: 205-500-4
FN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Indeks: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 5
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CH₃COOC₂H₅) (by GC, corrected for water) ≥99.6 %
Titrable Acid (µeq/g) ≤0.8
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, volumetric) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR