Diklormetan ≥99.8% stabilised, ULTRA RESI-ANALYZED® for analyse av organiskrester, J.T.Baker®

Leverandør: Avantor
Warning


Stabilisering: Stabilised with a methanol-alkene preservative system
Packaged under inert gas.

9264.1000EA 1825 NOK
9264.1000 9264.2500
Diklormetan ≥99.8% stabilised, ULTRA RESI-ANALYZED® for analyse av organiskrester, J.T.Baker®
Diklormetan
Formel: CH₂Cl₂
Kokepunkt: 39,8 °C (1013 hPa)
Smeltepunkt: –95 °C
Tetthet: 1,322 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
FN: 1593
ADR: 6.1,III
Merck Indeks: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 5
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CH₂Cl₂) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Chloride (Cl) ≤10 ppm
Water (by KF, coulometric) ≤0.02 %
Contains a methanol-alkene preservative system.

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR