Petroleumbensin, 30…60 °C, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Leverandør: Avantor
Danger


Packaged under inert gas.

9265.1000EA 1350 NOK
9265.1000 9265.2500
Petroleumbensin, 30…60 °C, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Petroleumbensin
Tetthet: ~0,68 kg (20 °C)
Flash punkt: −18 °C
MDL nummer: MFCD00081849
CAS-nummer: 8032-32-4
EINECS: 265-151-9
FN: 1268
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 5
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤ 5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤ 10
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤ 5
Color (APHA) ≤ 10
Residue after Evaporation ≤ 5 ppm
Water (by KF, coulometric) ≤ 0.05 %
Boiling Range(Lower) 30 - 60 °C
Boiling Range(Upper) 30 - 60 °C
ECD-Sensitive Impurities (as Ethylene Dibromide)
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR