VWR®, medietilskudd for dehydrerte kulturmedier
Leverandør: VWR Chemicals
602730ZLEA
2300
NOK
602730ZL
968130NL
968290NL
928010NL
928020NL
928030NL
928090NL
928110OL
928140NL
928150NL
928170OL
928180NL
928190NL
928200NL
928230NL
928240OL
928250NL
928260NL
928280NL
928320NL
928330NL
928350NL
928360NL
928370NL
928380NL
928390NL
928300NL
928340NL
303570ZK
335032ZF
928460NL
928080NL
928240NL
355070ZF
928620NL
928860NL
84721.0002
928050NL
928510NL
VWR®, medietilskudd for dehydrerte kulturmedier
Mikrobiologiske medier
Mikrobiologiske tilsetninger og supplementer
These standard supplements can be used with several culture media to prepare the complete culture media for your specific purpose. Most of these supplements are available as freeze-dried in vials and this ensures.
- No toxic agent aerosols released upon opening
- Standard volumes make preparation easier
- User friendly screw caps
- Can be stored at room temperature
SPESIFIKASJONER
Environmentally Preferable |