VWR®, medietilskudd for dehydrerte kulturmedier
Leverandør: VWR Chemicals
602730ZLEA 2300 NOK
602730ZL 968130NL 968290NL 928010NL 928020NL 928030NL 928090NL 928110OL 928140NL 928150NL 928170OL 928180NL 928190NL 928200NL 928230NL 928240OL 928250NL 928260NL 928280NL 928320NL 928330NL 928350NL 928360NL 928370NL 928380NL 928390NL 928300NL 928340NL 303570ZK 335032ZF 928460NL 928080NL 928240NL 355070ZF 928620NL 928860NL 84721.0002 928050NL 928510NL
VWR®, medietilskudd for dehydrerte kulturmedier
Mikrobiologiske medier Mikrobiologiske tilsetninger og supplementer
These standard supplements can be used with several culture media to prepare the complete culture media for your specific purpose. Most of these supplements are available as freeze-dried in vials and this ensures.
  • No toxic agent aerosols released upon opening
  • Standard volumes make preparation easier
  • User friendly screw caps
  • Can be stored at room temperature