Multi-cation standard 2 for IC, TraceCERT®, Supelco®
Leverandør: Merck
93159-100ML
93159-100MLEA 2750 NOK
93159-100ML
Multi-cation standard 2 for IC, TraceCERT®, Supelco®
Standarder Kationstandarder Multi-Element Standards
Ba, 100 mg/l; Ca, 100 mg/l; K, 100 mg/l
Li, 100 mg/l; Mg, 100 mg/l; Mn, 100 mg/l
Na, 100 mg/l; NH4, 100 mg/l; Sr, 100 mg/l
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034