Tris(2-ethylhexyl)phosphate, Supelco®

Leverandør: Merck

Synonymer: Trioctyl phosphate, Phosphoric acid tris(2-ethylhexyl ester), Tris(2-ethylhexyl) phosphate

MQ100

93299-25ML
93299-25MLEA 3425 NOK
93299-25ML
Tris(2-ethylhexyl)phosphate, Supelco®
Tris(2-ethylhexyl)phosphate
Formel: C₂₄H₅₁O₄P
MW: 434.64 g/mol
MDL nummer: MFCD00009491
CAS-nummer: 78-42-2
EINECS: 201-116-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1d728cd378a5a163bc24406de23bd35f1d59867&vid=89fca17d7f69e1f6f5623eb7813a67039b763f13&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR