n-Pentan ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: Pentane

Packaged under inert gas.

9333-03EA 26325 NOK
9333-03
n-Pentan ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
n-Pentane
Formel: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kokepunkt: 35…36 °C (1013 hPa)
Smeltepunkt: –130 °C
Tetthet: 0.63 g/cm³ (20 °C)
Flash punkt: –40 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009498
CAS-nummer: 109-66-0
EINECS: 203-692-4
FN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Indeks: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 5
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤5
Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR