Organic reference standard, Ethylhexyl triazone, analytical standard, Supelco®
Leverandør: Merck
93465-100MG
93465-100MGEA 1025 NOK
93465-100MG
Organic reference standard, Ethylhexyl triazone, analytical standard, Supelco®
Standarder Organiske standarder
Ethylhexyl triazone is a ultraviolet-B filter, which is used as a sunscreen agent in skin care products.
Forpakning: Bottomless glass bottles are inserted with fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65496fe094e07972a83e0f9ccf4edaf53a85a0aa&vid=ed00085c6622e4c6605508f21ef376a6ada5012f&prid=web&clid=SAPDAM