Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Leverandør: Merck
93468-100ML
93468-100MLEA 3670 NOK
93468-100ML
Transition metal mix 3 for ICP, 7 elements, TraceCERT®, Supelco®
Standarder Kationstandarder Multi-Element Standards
100 mg/l each: Ru, Rh, Pd, Os, Ir, Pt and Au in 10% hydrochloric acid
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034