Supelco® TraceCERT® Organic Reference Standard, Ethirimol

Leverandør: Merck

93638-50MG
93638-50MGEA 1050 NOK
93638-50MG
Supelco® TraceCERT® Organic Reference Standard, Ethirimol
Standarder Organiske standarder

Ethirimol is the ethylamino isomer of dimethirimol that belongs to a group of pyrimidine fungicides. This systemic fungicide acts by inhibiting nucleic acid synthesis and is used as a seed treatment to control powdery mildew in spring barley and wheat. Ethirimol is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. But a default maximum residue level (MRLs) from 0,01 to 2 mg/kg is applied for products of plant and animal origin according to Regulation (EU) 2021/1531. Ethirimol has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP) run within the EU and EFTA in/on products of plant origin.


Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


NB: P280 - P302 + P352 + P312 - P362 + P364 - P501

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 23947-60-6
MDL No. MFCD00055519
Molekylvekt 209,29
Hardhetsklasse beskrivelse H312
Lagringsforhold 15...25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR