Pentafluorophenylhydrazine ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®

Leverandør: Merck
Warning

93742-1G
93742-1GEA 190 NOK
93742-1G
Pentafluorophenylhydrazine ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Pentafluorophenylhydrazine

Pentafluorophenylhydrazine may be used as a derivatizing agent in the determination of semi-volatile carbonyl compounds in air and human plasma samples, using high-resolution gas chromatography coupled with gas chromatography (HRGC-MS) and liquid chromatography atmospheric-pressure chemical-ionization mass spectrometry (LC-APCI-MS-MS), respectively.

Formel: C₆H₃F₅N₂
MW: 198.1 g/mol
MDL nummer: MFCD00007574
CAS-nummer: 828-73-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR