Ortofosforsyre 49,5 - 50,5%, LiChropur™ for HPLC, Supelco®

Leverandør: Merck
Danger

Synonymer: Phosphoric acid

93752-250ML
93752-250MLEA 835 NOK
93752-250ML
Ortofosforsyre 49,5 - 50,5%, LiChropur™ for HPLC, Supelco®
Ortofosforsyre
Formel: H₃PO₄
MW: 98 g/mol
Kokepunkt: 158
Smeltepunkt: 21
Tetthet: 1.331...1.336 g/cm³ (20 °C)
MDL nummer: MFCD00011340
CAS-nummer: 7664-38-2
FN: 1805
ADR: 8 III
Merck Indeks: 14,07344

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) Colorless
Appearance (Form) Liquid
Titration (T) NaOH 1M 49.5 - 50.5 %
Suitability Test (HPLC) Corresponds to Standard
Density D20/4 1.331 - 1.336
UV-Absorption 230nm ≤ 0.03, 230nm ≤ 0.03
Test by UV Spectroscopy 240nm ≤ 0.02, 280nm ≤ 0.02, 500nm ≤ 0.02
Residue (Filter Test) No Residue
Filtertest (Method) Neat
Recommended Retest Period 54 Months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR