Supelco® Organic Reference Standard, Cyanidin chloride

Leverandør: Merck

94099-10MG
94099-10MGEA 2850 NOK
94099-10MG
Supelco® Organic Reference Standard, Cyanidin chloride
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9afa90c2035bbbdb09fe7ce3c72918aaedb2be0a&vid=9afa90c2035bbbdb09fe7ce3c72918aaedb2be0a&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 3,3′,4,5,7-Pentahydroxyflavylium chloride; Cyanidol chloride
M-Clarity quality level MQ100
CAS-nr 528-58-5
MDL No. MFCD00017582
Molekylvekt 322,70
Fraktforhold STD
Lagringsforhold 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR