VWR® Catalase Reagent
Leverandør: VWR Chemicals
941970ZAEA 400 NOK
941970ZA
VWR® Catalase Reagent
Mikrobiologiske testsystemer Enzymatiske tester
The catalase test can differentiate or identify microorganisms by checking their ability to produce catalase, an enzyme that protects microorganisms from toxic oxygen metabolites. Usually only aerobic microorganisms produce catalase.
  • A simple test to identify aerotolerant strains
  • Allows identification of Gram-positive cocci
  • Easy to use
  • Storage: 8 to 25 °C