Dimetylsulfoksid, Supelco®

Leverandør: Merck

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

M-Clarity™ quality level = MQ100

94563-10ML-F 94563-1ML-F
94563-10ML-FEA 4575 NOK
94563-10ML-F 94563-1ML-F
Dimetylsulfoksid, Supelco®
Dimetylsulfoksid
Formel: (CH₃)₂SO
Kokepunkt: 189 °C (1013 hPa)
Tetthet: 1,101 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4c9ae5d2d3449574cd07e8cf3d6b98bf6b21574&vid=5c5fa1b189adecb2e0f85132e3bbd52b9a32031f&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR