Trifluoreddiksyre ≥99.5%, BAKER ANALYZED® HPLC, J.T.Baker®

Leverandør: Avantor

Synonymer: TFA

9470.0010EA 2100 NOK
9470.0010
Trifluoreddiksyre ≥99.5%, BAKER ANALYZED® HPLC, J.T.Baker®
Trifluoreddiksyre

Gradient tested for HPLC Peptide and Protein Analysis

Formel: C₂HF₃O₂
MDL nummer: MFCD00004169
CAS-nummer: 76-05-1
FN: 2699
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay min. 99.5 %
Heavy Metals (as Pb) max. 10 ppm
Residue after Evaporation max. 10 ppm
Gradient Elution Test (a.u.)
at 215 nm max. 0.05
Trace Impurities (in ppm)
Iron (Fe) max. 5
Ultraviolet Absorbance (1.00-cm path vs water):
at 230 nm max. 0.15
at 254 nm max. 0.01
UV Cut-off, nm max. 210


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR