Hydrogen ionophore II, Selectophore™ ionophore, Supelco®

Leverandør: Merck

Synonymer: ETH 1907, 4-Nonadecylpyridine, Proton ionophore II

M-Clarity™ quality level = MQ100

95295-50MG
95295-50MGEA 4025 NOK
95295-50MG
Hydrogen ionophore II, Selectophore™ ionophore, Supelco®
Hydrogen ionophore II

H+-ionophore showing a linear response from pH 2 to 9 in an optimized membrane composition.


Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₄H₄₃N
MW: 345.60 g/mol
MDL nummer: MFCD00144460
CAS-nummer: 70268-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e302623fd99360718748ce4a1eb8dc2d67582fb4&vid=81b5aa8d6b256ec2cf29ac093902d54d210ecce1&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR