Organic reference standard, (+/-)-1-Amino-2-propanol, analytical standard, Supelco®

Leverandør: Merck

95343-500MG
95343-500MGEA 635 NOK
95343-500MG
Organic reference standard, (+/-)-1-Amino-2-propanol, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a01db7882ed843ea54a718cfbc66c7980c275733&vid=a01db7882ed843ea54a718cfbc66c7980c275733&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse (+/-)-1-Amino-2-propanol
Environmentally Preferable
CAS-nr 78-96-6
Formel C₃H₉NO 
Smeltepunkt −2 °C (lit.)
kokepunkt 160 °C (lit.)
Lagringstemp 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR