o-Xylen ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Leverandør: Honeywell Chemicals
Danger

Synonymer: 1,2-Dimethylbenzene

95662-250ML 95662-2.5L 95662-1L
95662-250MLEA 565 NOK
95662-250ML 95662-2.5L 95662-1L
o-Xylen ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
o-Xylen

Beilstein Number: 1815558

Formel: C₈H₁₀
MW: 106.17 g/mol
Kokepunkt: 7.6 %(V)
Smeltepunkt: –25 °C
Tetthet: 0.88 g/cm³ (20 °C)
Flash punkt: 30 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008519
CAS-nummer: 95-47-6
EINECS: 202-422-2
FN: 1307
ADR: 3 III
Merck Indeks: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.504 - 1.506
Non-volatile matter Max. 0.002 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
m-, p-Xylene (GC) Max. 0.6 %
Toluene (GC) Max. 0.4 %
Explosion Limit 7 %
Vapor Density 3.7 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR