Saltsyre 30%, BAKER INSTRA-ANALYZED®, J.T.Baker®

Leverandør: Avantor
Danger

9600.1000PEEA 1800 NOK
9600.1000PE
Saltsyre 30%, BAKER INSTRA-ANALYZED®, J.T.Baker®
Saltsyre
Formel: HCl
Kokepunkt: 110 °C (1013 hPa)
Smeltepunkt: –30 °C
Tetthet: 1,18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Assay (w/w) 29 - 31 %
Colour (Hazen) max. 10
Trace Impurities (in ppm)
Ammonium (NH4) max. 0.5
Bromide (Br) max. 10
Free Chlorine max. 0.5
Total Phosphorus max. 0.01
Total Sulfur max. 0.3
Trace Impurities (in ppb)
Silver (Ag) max. 1
Aluminum (Al) max. 1
Arsenic (As) max. 0.5
Gold (Au) max. 0.5
Boron (B) max. 1
Barium (Ba) max. 0.1
Beryllium (Be) max. 0.1
Bismuth (Bi) max. 0.1
Calcium (Ca) max. 1
Cadmium (Cd) max. 0.1
Cerium (Ce) max. 0.1
Cobalt (Co) max. 0.1
Chromium (Cr) max. 0.5
Cesium (Cs) max. 0.1
Copper (Cu) max. 0.5
Dysprosium (Dy) max. 0.1
Erbium (Er) max. 0.1
Europium (Eu) max. 0.1
Iron (Fe) max. 1
Gallium (Ga) max. 0.1
Gadolinium (Gd) max. 0.1
Germanium (Ge) max. 1
Hafnium (Hf) max. 0.1
Mercury (Hg) max. 0.1
Holmium (Ho) max. 0.1
Indium (In) max. 0.1
Potassium (K) max. 1
Lanthanum (La) max. 0.1
Lithium (Li) max. 0.1
Lutetium (Lu) max. 0.1
Magnesium (Mg) max. 0.5
Manganese (Mn) max. 0.1
Molybdenum (Mo) max. 0.1
Sodium (Na) max. 1
Niobium (Nb) max. 0.1
Neodymium (Nd) max. 0.1
Nickel (Ni) max. 0.5
Lead (Pb) max. 0.1
Palladium (Pd) For Information Only
Praseodymium (Pr) max. 0.1
Platinum (Pt) For Information Only
Rubidium (Rb) max. 0.1
Rhenium (Re) max. 0.1
Rhodium (Rh) max. 0.1
Ruthenium (Ru) max. 0.1
Antimony (Sb) max. 0.5
Scandium (Sc) max. 0.1
Selenium (Se) max. 1
Samarium (Sm) max. 0.1
Tin (Sn) max. 0.5
Strontium (Sr) max. 0.1
Tantalum (Ta) For Information Only
Terbium (Tb) max. 0.1
Tellurium (Te) max. 0.1
Thorium (Th) max. 0.1
Titanium (Ti) max. 0.5
Thallium (Tl) max. 0.1
Thulium (Tm) max. 0.1
Uranium (U) max. 0.1
Vanadium (V) max. 0.5
Tungsten (W) max. 0.1
Yttrium (Y) max. 0.1
Ytterbium (Yb) max. 0.1
Zinc (Zn) max. 1
Zirconium (Zr) max. 0.1

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR