Supelco® TraceCERT® Organic Reference Standard, Tris(2-chloroethyl) phosphate

Leverandør: Merck

96382-100MG
96382-100MGEA 740 NOK
96382-100MG
Supelco® TraceCERT® Organic Reference Standard, Tris(2-chloroethyl) phosphate
Standarder Organiske standarder

  • GHS signal: GHS08, GHS07, GHS09


Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


NB: P201 - P301 + P312 + P330 - P308 + P313

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf36c239a6b698edbd5b0c2134312a7e0f244fd6&vid=cf36c239a6b698edbd5b0c2134312a7e0f244fd6&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

M-Clarity quality level MQ300
Synonym(s) Phosphoric acid tris(2-chloroethyl) ester
Environmentally Preferable
Tetthet 1,39 g/ml at 25 °C (lit.)
CAS-nr 115-96-8
MDL No. MFCD00000967
Formel (ClCH₂CH₂O)₃P(O)
Molekylvekt 285,49
kokepunkt 192 °C/10 mmHg (lit.)
Hardhetsklasse beskrivelse H302 - H351 - H360
Flammepunkt 232 °C - Closed cup
Fraktforhold STD
Lagringsforhold 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR