Zincon sodium salt for spectrophotometric determination of Cu, for the determination of zinc, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

96440-1G 96440-5G
96440-1GEA 925 NOK
96440-1G 96440-5G
Zincon sodium salt for spectrophotometric determination of Cu, for the determination of zinc, Supelco®
Zincon sodium salt

A colorimetric indicator for Zn, Cu and Hg

Formel: C₂₀H₁₅N₄NaO₆S
MDL nummer: MFCD00064385
CAS-nummer: 62625-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf7e97635cfe147a06630f44f72026009afd0a78&vid=3990d3db741b4a8201c2ea104695f54d471d7b57&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8b39b66384f1ff15595062049c6f7f6a2aad621&vid=c9d2c1b4dd0c07ef04dd974489eacaa9636633d6&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR