Perchloric acid 69 - 72%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®

Leverandør: Avantor
Danger

9653-00EA 3525 NOK
9653-00
Perchloric acid 69 - 72%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Perchloric acid
Formel: HClO₄
MW: 100.46 g/mol
Kokepunkt: 160…181 °C (1013 hPa)
Smeltepunkt: –112.2 °C
Tetthet: 1.53…1.768 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
FN: 1873
ADR: 5.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use 69.0 - 72.0 %
ACS - Assay (HClO₄) 69.0 - 72.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤10 ppm
Chloride (Cl) ≤10 ppm
Nitrogen Compounds (as N) ≤10 ppm
ACS - Silicate and Phosphate (as SiO₂) ≤5 ppm
Sulfate (SO₄) ≤10 ppm
Trace Impurities - Aluminum (Al) ≤0.050 ppm
Trace Impurities - Barium (Ba) ≤0.100 ppm
Trace Impurities - Cadmium (Cd) ≤0.005 ppm
Trace Impurities - Calcium (Ca) ≤1 ppm
Trace Impurities - Chromium (Cr) ≤0.050 ppm
Trace Impurities - Cobalt (Co) ≤0.005 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Heavy Metals (as Pb) ≤0.1 ppm
Trace Impurities - Iron (Fe) ≤0.050 ppm
Trace Impurities - Lead (Pb) ≤0.010 ppm
Trace Impurities - Lithium (Li) ≤0.100 ppm
Trace Impurities - Magnesium (Mg) ≤0.050 ppm
Trace Impurities - Manganese (Mn) ≤0.005 ppm
Trace Impurities - Mercury (Hg) ≤0.005 ppm
Trace Impurities - Nickel (Ni) ≤0.005 ppm
Trace Impurities - Potassium (K) ≤0.500 ppm
Trace Impurities - Silicon (Si) ≤0.100 ppm
Trace Impurities - Silver (Ag) ≤0.005 ppm
Trace Impurities - Sodium (Na) ≤1.000 ppm
Trace Impurities - Strontium (Sr) ≤0.020 ppm
Trace Impurities - Tin (Sn) ≤0.010 ppm
Trace Impurities - Zinc (Zn) ≤0.050 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR