Svovelsyre 95.0-98.0% (by acidimetry) NF, ACS, J.T.Baker®

Leverandør: Avantor
Danger

9661-03EA 0 NOK
9661-03
Svovelsyre 95.0-98.0% (by acidimetry) NF, ACS, J.T.Baker®
Svovelsyre
Formel: H₂SO₄
MW: 98.08 g/mol
Kokepunkt: 330
Smeltepunkt: 10.38
Tetthet: 1.84
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
FN: 1830
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product
Meets N.F. Requirements
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
NF - Arsenic (As) ≤1 ppm
NF - Chloride (Cl) ≤0.005 %
NF - Identification Passes Test
NF - Reducing Substances Passes Test
NF - Residue on Ignition ≤0.005 %
Ammonium (NH₄) ≤1 ppm
Appearance Passes Test
Assay (H₂SO₄) 95.0 - 98.0 %
Chloride (Cl) ≤0.1 ppm
Color (APHA) ≤10
Heavy Metals (as Pb) ≤500.0 ppb
Nitrate (NO₃) ≤0.5 ppm
Residue after Ignition ≤4 ppm
Specific Gravity at 60°/60°F ≥1.84
Substances Reducing Permanganate (as SO₂) ≤2 ppm
Trace Impurities - Aluminum (Al) ≤200.0 ppb
Trace Impurities - Arsenic (As) ≤0.004 ppm
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gold (Au) ≤100.0 ppb
Trace Impurities - Iron (Fe) ≤200.0 ppb
Trace Impurities - Lead (Pb) ≤200.0 ppb
Trace Impurities - Magnesium (Mg) ≤200.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤200.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Tin (Sn) ≤200.0 ppb
Trace Impurities - Titanium (Ti) ≤200.0 ppb
Trace Impurities - Zinc (Zn) ≤200.0 ppb
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR