N,N-Dimetylformamid ≥99.9%, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®

Leverandør: Avantor

Synonymer: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

9753.1000EA 2300 NOK
9753.1000
N,N-Dimetylformamid ≥99.9%, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
N,N-Dimetylformamid
Formel: HCON(CH₃)₂
MW: 73.09 g/mol
Kokepunkt: 153
Smeltepunkt: -61
Flash punkt: 57.5
MDL nummer: MFCD00003284
CAS-nummer: 68-12-2
FN: 2265
ADR: 3 III
Merck Indeks: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (by GC) min. 99.9 %
Water (by KF) max. 0.03 %
Refractive Index n20/D Dependent on chemical
Acidity max. 0.0005 meq/g
Residue after Evaporation max. 0.0005 %
GC-HS Suitability passes test
1 Class solvents residue
Benzene indeterminable
Carbon tetrachloride indeterminable
1,2-Dichloroethane indeterminable
1,1-Dichloroethane indeterminable
1,1,1-Trichloroethane indeterminable
2 Class solvents residue
Acetonitrile max. 4 ppm
Cyclohexane max. 1 ppm
Dichloromethane max. 0.6 ppm
1,2-Dimethoxyethane max. 5 ppm
1,4-Dioxane max. 4 ppm
Hexane max. 0.3 ppm
Methanol max. 1 ppm
Methylcyclohexane max. 1 ppm
Pyridine max. 1 ppm
Tetrahydrofuran max. 0.7 ppm
Toluene max. 0.9 ppm
p-xylen max. 1 ppm
m-Xylen max. 1 ppm
o-xylene max. 1 ppm
3 Class solvents residue in total: max. 0.5 %
Acetone max. 1 ppm
1-Butanol max. 1 ppm
Butyl acetate max. 1 ppm
Ethanol max. 1 ppm
Ethyl acetale max. 1 ppm
Heptane max. 1 ppm
Iso-propyl acetate max. 1 ppm
1-Propanol max. 1 ppm
2-Propanol max. 1 ppm
Suitable as solvent for the analysis of residual solvents of ICH classes 1,2 and 3 according to Ph.Eur. and USP.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR