Natriumformat solution_SC 10 mM NaOH 2-propanol/water (1:1) (+0.2% HCOOH) LC-MS, Fluka™

Leverandør: Honeywell Chemicals
Danger

Synonymer: Formic acid sodium salt

97574-100ML
97574-100MLEA 2625 NOK
97574-100ML
Natriumformat solution_SC 10 mM NaOH 2-propanol/water (1:1) (+0.2% HCOOH) LC-MS, Fluka™
Natriumformat
Formel: HCO₂Na
MW: 68.01 g/mol
Smeltepunkt: 257.31
Flash punkt: 22 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013101
CAS-nummer: 141-53-7
FN: 1219
ADR: 3 II
Merck Indeks: 13,08694

Bestill nå

Spesifikasjon Testresultater

Identity (LC-MS) complying
LC-MS application complying
Suitability for the LC-MS complying

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR