2-Merkaptoetanol ≥99.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®

Leverandør: Merck
Danger

Synonymer: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

97622-10ML 97622-1ML 97622-10X1ML
97622-10MLEA 478 NOK
97622-10ML 97622-1ML 97622-10X1ML
2-Merkaptoetanol ≥99.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
2-Merkaptoetanol

Suitable for derivatisation of amine groups.

Formel: HSCH₂CH₂OH
MW: 78.14 g/mol
Kokepunkt: 157
Smeltepunkt: -100
Flash punkt: 68
MDL nummer: MFCD00004890
CAS-nummer: 60-24-2
FN: 2966
ADR: 6.1 II
Merck Indeks: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50a0997da05082d8384dbdad2b0f714fa5823da3&vid=940b400110902081cb91993d15b274ca3aea6670&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR