Natriumhydroksid 2 mol/l (2 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Danger

98108.290EA 515 NOK
98108.290 98108.360 98108.400
Natriumhydroksid 2 mol/l (2 N) in aqueous solution, AVS TITRINORM volumetric solution
Natriumhydroksid
Formel: NaOH
Kokepunkt: 109 °C (1013 hPa)
Tetthet: 1,078 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 1.996 - 2.004 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR