J.T.Baker®, Cycle-Tap Sampler
Leverandør: Avantor
9815-05EA 0 NOK
9815-05
J.T.Baker®, Cycle-Tap Sampler
Prøvetakere Væskeprøvetakere
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.